missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 10377264
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10377264 5 g 5g
10452864 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10377264 Supplier Thermo Scientific Maybridge Supplier No. CC52416DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-34-0
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD09025875
InChI Key ITXQSYHCKPGZLV-UHFFFAOYSA-N
Synonym 4-tetrahydropyran-4-yloxy benzonitrile, 4-oxan-4-yloxy benzonitrile, 4-tetrahydro-2h-pyran-4-yloxy benzonitrile, 4-oxan-4-yl oxy benzonitrile, 4-tetrahydro-2h-pyran-4-yl oxy benzonitrile, benzonitrile,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzenecarbonitrile
PubChem CID 24229527
IUPAC Name 4-(oxan-4-yloxy)benzonitrile
SMILES C1COCCC1OC2=CC=C(C=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 4-(Tetrahydropyran-4-yloxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.