missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Pentylphenol, 99%

Product Code. 11499986
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499986 50 g 50g
11409996 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499986 Supplier Thermo Scientific Alfa Aesar Supplier No. B20939.18
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 80-46-6 | C11H16O | 164.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-46-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002369
InChI Key NRZWYNLTFLDQQX-UHFFFAOYSA-N
Synonym 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
PubChem CID 6643
ChEBI CHEBI:35096
IUPAC Name 4-(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=C(C=C1)O

Specifications

Melting Point 88°C to 89°C
Boiling Point 254°C to 256°C
Quantity 50 g
UN Number UN2430
Merck Index 14,7142
Formula Weight 164.25
Percent Purity 99%
Chemical Name or Material 4-tert-Pentylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.