missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Octylphenol Diethoxylate, TRC

Product Code. 30411923
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411923 25 mg
30419149 5 mg
30417847 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411923 Supplier Toronto Research Chemicals Supplier No. TRCO29379525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2315-61-9
Molecular Formula C18 H30 O3
Molecular Weight (g/mol) 294.43
Synonym Ethanol, 2-[2-[p-(1,1,3,3-tetramethylbutyl)phenoxy]ethoxy]- (7CI,8CI), 2-[2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]ethoxy]ethanol, 2-[2-(p-Octylphenoxy)ethoxy]ethanol, 4-Octylphenol diethoxylate
IUPAC Name 2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethanol
SMILES CC(C)(C)CC(C)(C)c1ccc(OCCOCCO)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H30O3/c1-17(2,3)14-18(4,5)15-6-8-16(9-7-15)21-13-12-20-11-10-19/h6-9,19H,10-14H2,1-5H3
Formula Weight 294.2195
Percent Purity >95%
Chemical Name or Material 4-tert-Octylphenol Diethoxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.