missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Octylphenol 13C6 (phenyl 13C6), Dr. Ehrenstorfer

Product Code. 30319727
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319727 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319727 Supplier Dr. Ehrenstorfer Supplier No. DREC15712110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-tert-Octylphenol ring-13C6
CAS 1173020-24-0
Molecular Formula 13C6 C8 H22 O
Quantity 10 mg
InChI Formula InChI=1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3/i6+1,7+1,8+1,9+1,11+1,12+1
SMILES CC(C)(C)CC(C)(C)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
Molecular Weight (g/mol) 212.28
Formula Weight 212.1872 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.