missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyltoluene, 96%

Product Code. 10053550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053550 25 mL Glass bottle 25mL
10440531 1 L Glass bottle 1 L
10701535 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10053550 Supplier Thermo Scientific Acros Supplier No. 296320250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-51-1 | C11H16 | 148.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-51-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00008837
InChI Key QCWXDVFBZVHKLV-UHFFFAOYSA-N
Synonym 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene
PubChem CID 7390
IUPAC Name 1-tert-butyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C(C)(C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -54.0°C
Color Colorless to Yellow
Density 0.8612g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4CH3
Refractive Index 1.4910 to 1.4930
Quantity 25 mL
Beilstein 05, 439
Specific Gravity 0.8612
Solubility Information Solubility in water: 0.06 wt% (20°C). Other solubilities: miscible with isopropanol,toluene and cyclohexane,soluble in nearly all organic solvents
Viscosity 1.25 mPa.s (25°C)
Formula Weight 148.25
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-tert-Butyltoluene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butyltoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.