missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol

Product Code. 11318765
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318765 100 mL 100mL
11328765 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318765 Supplier Thermo Scientific Alfa Aesar Supplier No. L12356.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1746-23-2 | C12H16 | 160.26 g/mol

4-tert-Butylstyrene is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1746-23-2
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00065126
InChI Key QEDJMOONZLUIMC-UHFFFAOYSA-N
Synonym 4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
PubChem CID 15627
IUPAC Name 1-tert-butyl-4-ethenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C=C

Specifications

Melting Point -38°C
Density 0.884
Boiling Point 219°C
Flash Point 80°C (176°F)
Refractive Index 1.525
Quantity 100 mL
Beilstein 1903714
Solubility Information Not miscible in water.
Formula Weight 160.26
Percent Purity 94%
Chemical Name or Material 4-tert-Butylstyrene, stabilized with 50ppm 4-tert-butylcatechol
Show More Show Less
compliance-icons
Product Identifier
  • 4-tert-Butylstyrene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-tert-Butylstyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.