missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylacetylene, 96%

Product Code. 10157882 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157882 25 g Glass bottle 25g
10200413 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157882 Supplier Thermo Scientific Acros Supplier No. 380850250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 772-38-3 | C12H14 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-38-3
Molecular Formula C12H14
Molecular Weight (g/mol) 158.24
MDL Number MFCD00190197
InChI Key ZSYQVVKVKBVHIL-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
PubChem CID 2757302
IUPAC Name 1-tert-butyl-4-ethynylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown-Khaki to Green
Boiling Point 130.0°C (40.0 mmHg)
Flash Point 82.2°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH≡CC6H4C(CH3)3
Refractive Index 1.5280 to 1.5300
Quantity 25 g
Formula Weight 158.24
Percent Purity 96%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-tert-Butylphenylacetylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenylacetylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.