missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylacetylene, 90+%

Product Code. 11344968
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344968 10 g 10g
11334968 2 g 2g
11324968 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344968 Supplier Thermo Scientific Alfa Aesar Supplier No. 043920.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 772-38-3 | C12H14 | 158.244 g/mol

4-tert-Butylphenylacetylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-38-3
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00190197
InChI Key ZSYQVVKVKBVHIL-UHFFFAOYSA-N
Synonym 1-tert-butyl-4-ethynylbenzene, 4-tert-butyl phenylacetylene, 4-tert-butylphenylacetylene, benzene, 1-1,1-dimethylethyl-4-ethynyl, acmc-20amtx, 4-t-butylphenylacetylene, 4-tert-butylphenyl ethyne, 4-tert.butylphenylacetylen, 4-tert-butyl phenylethyne, 4-tert-butylphenyl acetylene
PubChem CID 2757302
IUPAC Name 1-tert-butyl-4-ethynylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C#C

Specifications

Density 0.878 g/mL
Boiling Point 130°C (40 mmHg)
Flash Point 82°C (180°F)
Odor Characteristic
Refractive Index 1.5297
Quantity 10 g
UN Number UN3082
Solubility Information Difficult to mix with water.
Formula Weight 158.24
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.