missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenoxyacetyl chloride, 98%

Product Code. 11444007
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11444007

Brand: Thermo Scientific Alfa Aesar A19408.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90734-55-7 | C12H15ClO2 | 226.70 g/mol

4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90734-55-7
Molecular Formula C12H15ClO2
Molecular Weight (g/mol) 226.70
MDL Number MFCD00798568
InChI Key CFTNMBDQWVPSHI-UHFFFAOYSA-N
Synonym 4-tert-butylphenoxyacetyl chloride, 4-tert-butylphenoxy acetyl chloride, 2-4-tert-butylphenoxy acetyl chloride, 2-4-tert-butyl phenoxy acetyl chloride, pubchem14851, 4-tert-butylphenoxyacetyl cloride, para-t-butylphenoxyacetyl chloride, 4-t-butyl-phenoxy acetyl chloride, 4-tert-butyl-phenoxyacetyl chloride
PubChem CID 611195
IUPAC Name 2-(4-tert-butylphenoxy)acetyl chloride
SMILES CC(C)(C)C1=CC=C(OCC(Cl)=O)C=C1

Specifications

Boiling Point 130°C to 132°C (5 mmHg)
Refractive Index 1.5165
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 226.71
Percent Purity 98%
Chemical Name or Material 4-tert-Butylphenoxyacetyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.