missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol, 99%

Product Code. 11462578
Change view
Click to view available options
Quantity:
500 g
2500 g
10 kg
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462578 10 kg 10000g
11442578 500 g 500g
11452578 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462578 Supplier Thermo Scientific Alfa Aesar Supplier No. A15871.0C
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-54-4 | C10H14O | 150.221 g/mol

4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

Solubility
Soluble in water. [8.7 g/L (20°C)]

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol

Specifications

Melting Point 97°C to 101°C
Density 0.908
Boiling Point 236°C to 238°C
Flash Point 113°C
Assay Percent Range 99%
Quantity 10 kg
UN Number UN3077
Beilstein 1817334
Merck Index 14,1585
Solubility Information Soluble aq. soln.. [8.7 g/L (20°C)]
SMILES CC(C)(C)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 150.22
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material 4-tert-Butylphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-tert-butylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.