missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyldimethylsilyloxybenzeneacetic Acid Methyl Ester, TRC

Product Code. 30331861
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331861 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331861 Supplier Toronto Research Chemicals Supplier No. TRCB691085250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-tert-Butyldimethylsilyloxybenzeneacetic Acid Methyl Ester
CAS 105460-59-1
Molecular Formula C15H24O3Si
Quantity 250 mg
Synonym Methyl 4-tert-Butyldimethylsilyloxyphenylacetate
InChI Formula InChI=1S/C15H24O3Si/c1-15(2,3)19(5,6)18-13-9-7-12(8-10-13)11-14(16)17-4/h7-10H,11H2,1-6H3
SMILES COC(=O)Cc1ccc(O[Si](C)(C)C(C)(C)C)cc1
IUPAC Name methyl 2-[4-[tert-butyl(dimethyl)silyl]oxyphenyl]acetate
Molecular Weight (g/mol) 280.43
Formula Weight 280.1495
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.