missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyldimethylsilyloxy)cyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 10234964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234964 5 g Glass bottle 5g
10133104 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234964 Supplier Thermo Scientific Acros Supplier No. 426850050
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55145-45-4
Molecular Formula C12H24O2Si
Molecular Weight (g/mol) 228.41
MDL Number MFCD06411307
InChI Key HXKBGMNGSYGPRB-UHFFFAOYSA-N
Synonym 4-tert-butyldimethylsilyloxy cyclohexanone, 4-tert-butyldimethylsilyl oxy cyclohexanone, 4-tert-butyl dimethyl silyl oxy cyclohexanone, 4-tert-butyl-dimethyl-silanyloxy-cyclohexanone, 4-tert-butyldimethylsiloxy cyclohexanone, 4-tert-butyldimethylsilyloxy cyclohexanone meso form, 4-1,1-dimethylethyl dimethylsilyl oxy cyclohexanone, cyclohexanone, 4-1,1-dimethylethyl dimethylsilyl oxy, 4-tert-butyl dimethyl silyl oxycyclohexanone
PubChem CID 10609430
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxycyclohexan-1-one
SMILES CC(C)(C)[Si](C)(C)OC1CCC(=O)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9090g/mL
Boiling Point 85°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (O=)C6H9OSi(CH3)2C(CH3)3
Quantity 5 g
Specific Gravity 0.909
Formula Weight 228.41
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(tert-Butyldimethylsilyloxy)cyclohexanone
Show More Show Less
compliance-icons
Product Identifier
  • 4-(tert-Butyldimethylsilyloxy)cyclohexanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.