missing translation for 'onlineSavingsMsg'
Learn More

4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde, 97%, Thermo Scientific™

Product Code. 10028189 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10028189 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10028189 Supplier Thermo Scientific Acros Supplier No. 428170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 120743-99-9
CAS Min % 96.0
CAS Max % 100.0
Density 0.9570g/mL
Boiling Point 224°C to 226°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C13H20O2Si
MDL Number MFCD05865174
Quantity 1 g
Specific Gravity 0.957
Synonym 4-t-butyldimethylsilyloxy benzaldehyde, 4-tert-butyldimethylsilyl oxy benzaldehyde, 4-tert-butyldimethylsilyloxy benzaldehyde, 4-t-butyldimethylsilyl oxy benzaldehyde, 4-tert-butyl dimethyl silyl oxy benzaldehyde, 4-tert-butyldimethylsiloxy benzaldehyde, acmc-209a8a
InChI Key XACWSBWCLJXKGI-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(C=O)C=C1
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxybenzaldehyde
Molecular Weight (g/mol) 236.39
PubChem CID 10879133
Formula Weight 236.39
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-[(tert-Butyldimethylsilyl)oxy]benzaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.