missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanone, 99%

Product Code. 11457487
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457487 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457487 Supplier Thermo Scientific Alfa Aesar Supplier No. A15612.22
  • ONLINE EXCLUSIVE PRICE
    £53.80
    £48.65 / 100g
    Save £5.15
    9% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-53-3 | C10H18O | 154.253 g/mol

4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylcyclohexanone is used as a perfuming agents and in cosmetic.

Solubility
Soluble in alcohol, ethanol (0.5g/10 mL). Insoluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-53-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001642
InChI Key YKFKEYKJGVSEIX-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanone, 4-tert-butyl cyclohexanone, p-tert-butylcyclohexanone, cyclohexanone, 4-1,1-dimethylethyl, cyclohexanone, p-tert-butyl, cyclohexanone, 4-tert-butyl, 4-t-butylcyclohexanone, 4-tert-butyl cyclohexan-1-one, .gamma.-tert-butylcyclohexanone, pubchem20244
PubChem CID 7392
IUPAC Name 4-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCC(=O)CC1

Specifications

Melting Point 47°C to 51°C
Density 0.893
Boiling Point 113°C to 116°C (20 mmHg)
Flash Point 96°C (204°F)
Quantity 100 g
Beilstein 507309
Solubility Information Soluble in alcohol,ethanol (0.5g/10 mL). Insoluble in water.
Formula Weight 154.25
Percent Purity 99%
Chemical Name or Material 4-tert-Butylcyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.