missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals

Catalog Number 10090580
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
98-52-2
C10H20O
156.27
MFCD00001473,MFCD00064952,MFCD00070476
CCOQPGVQAWPUPE-UHFFFAOYSA-N
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
7391
4-tert-butylcyclohexan-1-ol
CC(C)(C)C1CCC(O)CC1
This item is not returnable. View return policy
98-52-2
C10H20O
156.27
MFCD00001473,MFCD00064952,MFCD00070476
CCOQPGVQAWPUPE-UHFFFAOYSA-N
4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
7391
4-tert-butylcyclohexan-1-ol
CC(C)(C)C1CCC(O)CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 60.0°C to 69.0°C
Color White
Boiling Point 224.0°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3CC6H10OH
Refractive Index 1.447
Quantity 5 g
Beilstein 06, I, 18
Solubility Information Solubility in water: <1g/L water (20 c)
Viscosity 8.68 mPa.s (80°C)
Formula Weight 156.27
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material 4-tert-Butylcyclohexanol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'