missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[4]arene, 99%

Product Code. 11447074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447074 25 g 25g
11437074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447074 Supplier Thermo Scientific Alfa Aesar Supplier No. B21515.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol

It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

Solubility
Soluble in chloroform, benzene, toluene. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60705-62-6
Molecular Formula C44H56O4
Molecular Weight (g/mol) 648.93
MDL Number MFCD00066280
InChI Key NVKLTRSBZLYZHK-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
PubChem CID 335377
IUPAC Name 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1

Specifications

Melting Point 339°C to 344°C
Quantity 25 g
Beilstein 2033965
Solubility Information Soluble in chloroform,benzene,toluene. Insoluble in water.
Formula Weight 648.93
Percent Purity 99%
Chemical Name or Material 4-tert-Butylcalix[4]arene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.