missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzyl Bromide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-tert-Butylbenzyl Bromide |
| CAS | 18880-00-7 |
| Molecular Formula | C11H15Br |
| Quantity | 10 g |
| Synonym | α-Bromo-p-tert-butyl-toluene, 1-(Bromomethyl)-4-(1,1-dimethylethyl)benzene, 1-Bromomethyl-4-tert-butylbenzene, 1-tert-Butyl-4-(bromomethyl)benzene, 4-(1,1-Dimethylethyl)benzyl bromide, 4-tert-Butyl-1-bromomethylbenzene, 4-tert-Butylphenylmethyl Bromide, Bromo(4-tert-butylphenyl)methane, NSC 186287, p-tert-Butylbenzyl Bromide, α-Bromo-p-tert-butyltoluene |
| InChI Formula | InChI=1S/C11H15Br/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7H,8H2,1-3H3 |
| SMILES | CC(C)(C)c1ccc(CBr)cc1 |
| IUPAC Name | 1-(bromomethyl)-4-tert-butylbenzene |
| Molecular Weight (g/mol) | 227.14 |
| Formula Weight | 226.0357 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction