missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl Bromide, TRC

Product Code. 30339781
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30339781

Brand: Toronto Research Chemicals TRCB6924561G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-tert-Butylbenzyl Bromide
CAS 18880-00-7
Molecular Formula C11H15Br
Quantity 1 g
Synonym α-Bromo-p-tert-butyl-toluene, 1-(Bromomethyl)-4-(1,1-dimethylethyl)benzene, 1-Bromomethyl-4-tert-butylbenzene, 1-tert-Butyl-4-(bromomethyl)benzene, 4-(1,1-Dimethylethyl)benzyl bromide, 4-tert-Butyl-1-bromomethylbenzene, 4-tert-Butylphenylmethyl Bromide, Bromo(4-tert-butylphenyl)methane, NSC 186287, p-tert-Butylbenzyl Bromide, α-Bromo-p-tert-butyltoluene
InChI Formula InChI=1S/C11H15Br/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7H,8H2,1-3H3
SMILES CC(C)(C)c1ccc(CBr)cc1
IUPAC Name 1-(bromomethyl)-4-tert-butylbenzene
Molecular Weight (g/mol) 227.14
Formula Weight 226.0357
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.