missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzyl bromide, 97%

Product Code. 11431630
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431630 25 g 25g
11421630 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431630 Supplier Thermo Scientific Alfa Aesar Supplier No. H26991.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18880-00-7 | C11H15Br | 227.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18880-00-7
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00000180
InChI Key QZNQSIHCDAGZIA-UHFFFAOYSA-N
Synonym 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide
PubChem CID 87836
IUPAC Name 1-(bromomethyl)-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)CBr

Specifications

Melting Point 8°C to 12°C
Density 1.236
Boiling Point 93°C to 94°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5465
Quantity 25 g
UN Number UN3265
Beilstein 471674
Formula Weight 227.14
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.