missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoyl chloride, 98%

Product Code. 11476957
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476957 100 g 100g
11466957 25 g 25g
11486957 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476957 Supplier Thermo Scientific Alfa Aesar Supplier No. A14373.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol

4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9?-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9′-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1710-98-1
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000695
InChI Key WNLMYNASWOULQY-UHFFFAOYSA-N
PubChem CID 74372
IUPAC Name 4-tert-butylbenzoyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)C(Cl)=O

Specifications

Density 1.08
Boiling Point 125°C to 126°C (10 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.5351
Quantity 100 g
UN Number UN3265
Beilstein 775793
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 4-1,1-dimethylethyl, 4-tert-butylbenzoylchloride, p-t-butylbenzoyl chloride, p-tert-butylbenzoyl chloride, benzoyl chloride,4-1,1-dimethylethyl, 4-tert-butyl-benzoyl chloride, 4-tert-butyl benzoyl chloride, 4-t-butylbenzoyl chloride, acmc-1brh5, 4-tertbutylbenzoyl chloride
Solubility Information Reacts with water.
Formula Weight 196.68
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.