missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzophenone, 98%

Product Code. 11332506
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332506 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11332506 Supplier Thermo Scientific Alfa Aesar Supplier No. L06386.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22679-54-5 | C17H18O | 238.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22679-54-5
Molecular Formula C17H18O
Molecular Weight (g/mol) 238.33
MDL Number MFCD00017255
InChI Key DFYJCXSOGSYMAJ-UHFFFAOYSA-N
Synonym 4-tert-butylbenzophenone, 4-t-butyl benzophenone, 4-tert-butyl phenyl phenyl methanone, 4-tert-butyl-benzophenone, 4-tert-butylphenyl-phenylmethanone, 4-tert-butylphenyl phenyl methanone, methanone, 4-1,1-dimethylethyl phenyl phenyl, acmc-20aman, 4-tert.-butylbenzophenone
PubChem CID 89790
IUPAC Name (4-tert-butylphenyl)-phenylmethanone
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 38°C to 42°C
Boiling Point 168°C to 170°C (2 mmHg)
Flash Point 150°C (302°F)
Quantity 1 g
Beilstein 2100075
Formula Weight 238.32
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.