missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoic acid, 99%

Product Code. 11467846
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467846 250 g 250g
11457846 1000 g 1000g
11477846 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467846 Supplier Thermo Scientific Alfa Aesar Supplier No. A13997.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-73-7 | C11H14O2 | 178.23 g/mol

4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-73-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00002563
InChI Key KDVYCTOWXSLNNI-UHFFFAOYSA-N
Synonym 4-tert-butyl benzoic acid, tbba, p-tert-butylbenzoic acid, benzoic acid, 4-1,1-dimethylethyl, 4-t-butylbenzoic acid, benzoic acid, p-tert-butyl, 4-tert-butyl-benzoic acid, p-t-butylbenzoic acid, 4-tert-butylbenzoicacid, unii-43z7t3vn0r
PubChem CID 7403
ChEBI CHEBI:34443
IUPAC Name 4-tert-butylbenzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 164°C to 168°C
Boiling Point 280°C (decomposition)
Flash Point 180°C (356°F)
Quantity 250 g
UN Number UN2811
Beilstein 607545
Solubility Information Insoluble in water.
Formula Weight 178.23
Percent Purity ≥99%
Chemical Name or Material 4-tert-Butylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.