missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzaldehyde, 97%

Product Code. 11452388
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452388 100 g 100g
11432388 5 g 5g
11442388 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452388 Supplier Thermo Scientific Alfa Aesar Supplier No. A15740.22
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 939-97-9 | C11H14O | 162.23 g/mol

Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for preparing isoxazolyl penicillin derivatives. 4-tert-Butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (KI) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes. It is also an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds.

Solubility
Soluble in water.

Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

Density 0.97
Boiling Point 244°C to 246°C
Flash Point 101°C (213°F)
Refractive Index 1.5285
Quantity 100 g
UN Number UN3082
Beilstein 1906461
Sensitivity Air and light sensitive
Solubility Information Soluble in water.
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.