missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butylacetophenone, 98%

Product Code. 11490174
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490174 1 g 1g
11400184 5 g 5g
11410184 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490174 Supplier Thermo Scientific Alfa Aesar Supplier No. A13615.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 943-27-1 | C12H16O | 176.259 g/mol

4'-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-27-1
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00017256
InChI Key UYFJYGWNYQCHOB-UHFFFAOYSA-N
Synonym 4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanone
PubChem CID 13669
IUPAC Name 1-(4-tert-butylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 17°C to 18°C (dimorphic)
Density 0.964
Boiling Point 107°C to 108°C (5 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.52
Quantity 1 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material 4'-tert-Butylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.