missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzenesulfonamide, Mikromol™

Product Code. 17933476
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933476 100 mg 100mg
17992996 25 mg 25mg
30462831 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933476 Supplier Mikromol Supplier No. MM3369.02
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(tert-Butyl)benzenesulfonamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(tert-butyl)benzenesulfonamide
CAS 6292-59-7
Analyte or Component Names 4-(tert-butyl)benzenesulfonamide
Shipping Condition Room Temperature
Molecular Formula C10 H15 N O2 S
Certifications/Compliance ISO 17025
Synonym Benzenesulfonamide, 4-(1,1-dimethylethyl)-, Benzenesulfonamide, p-tert-butyl- (6CI,8CI), 4-(1,1-Dimethylethyl)benzenesulfonamide, 4-tert-Butylbenzenesulfonamide, NSC 9911, p-(tert-Butyl)phenylsulfonamide, p-tert-Butylbenzenesulfonamide, (4-(tert-butyl)benzenesulfonamide), Bosentan Related Compound E, Bosentan Related Compound E
InChI Formula InChI=1S/C10H15NO2S/c1-10(2,3)8-4-6-9(7-5-8)14(11,12)13/h4-7H,1-3H3,(H2,11,12,13)
SMILES CC(C)(C)c1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-tert-butylbenzenesulfonamide
Molecular Weight (g/mol) 213.3
Quantity 100 mg
Formula Weight 213.0823 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.