missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)benzene-1,2-diamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | (CH3)3CC6H3(NH2)2 |
Quantity | 1 g |
Synonym | 4-tert-butyl benzene-1,2-diamine, 4-tert-butyl-1,2-diaminobenzene, 1,2-diamino-4-tert-butylbenzene, 4-tert-butyl-o-phenylenediamine, 1,2-benzenediamine,4-1,1-dimethylethyl, 1,2-benzenediamine, 4-1,1-dimethylethyl, pubchem22919, 4-tert-butyl-diaminobenzene, 4-t-butylbenzene-1,2-diamine, 4-tert-butylbenzen-1,2-diamine |
Formula Weight | 164.25 |
Percent Purity | 97% |
Chemical Name or Material | 4-(tert-Butyl)benzene-1, 2-diamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'