missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)aniline, 97%, Thermo Scientific™

Product Code. 10356664
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356664 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10356664 Supplier Thermo Scientific Maybridge Supplier No. SB01038DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 769-92-6
CAS Min % 97.0
Density 0.93g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H15N
Linear Formula (CH3)3CC6H4NH2
MDL Number MFCD00007899
Quantity 1 g
Synonym 4-tert-butyl aniline, p-tert-butylaniline, benzenamine, 4-1,1-dimethylethyl, p-t-butylaniline, 4-t-butylaniline, unii-47h9440zow, 4-t-butylbenzeneamine, 4-tert-butylbenzenamine, 4-tert-butylbenzeneamine
InChI Key WRDWWAVNELMWAM-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC=C(C=C1)N
IUPAC Name 4-tert-butylaniline
Molecular Weight (g/mol) 149.237
PubChem CID 69861
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material 4-(Tert-butyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.