missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonyl)thiomorpholine-3-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10557464
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10557464 5 g 5g
10229292 250 mg 250mg
10790904 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10557464 Supplier Thermo Scientific Maybridge Supplier No. CC42401DE
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 128453-98-5
Molecular Formula C10H17NO4S
Molecular Weight (g/mol) 247.309
MDL Number MFCD02683067
InChI Key CTDIKDIZNAGMFK-UHFFFAOYSA-N
Synonym 4-tert-butoxycarbonyl thiomorpholine-3-carboxylic acid, 4-boc-thiomorpholine-3-carboxylic acid, n-boc-3-thiomorpholinecarboxylic acid, thiomorpholine-3,4-dicarboxylic acid 4-tert-butyl ester, 4-tert-butoxy carbonyl thiomorpholine-3-carboxylic acid, 3,4-thiomorpholinedicarboxylicacid, 4-1,1-dimethylethyl ester, acmc-20dpph, acmc-1bvvh, +/--thiomorpholine-3,4-dicarboxylicacid4-tert-butylester, n-boc-3-thiomorpholinecarboxylicacid
PubChem CID 2756831
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonyl]thiomorpholine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCSCC1C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 247.31
Percent Purity 97%
Chemical Name or Material 4-(Tert-butoxycarbonyl)thiomorpholine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.