missing translation for 'onlineSavingsMsg'
Learn More

4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10069591
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10069591 1 g Amber Glass Bottle 1g
10753673 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10069591 Supplier Thermo Scientific Maybridge Supplier No. SPB08449DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 77716-11-1
Molecular Formula C11H16N2O4
Molecular Weight (g/mol) 240.259
MDL Number MFCD02094544
InChI Key GOLAEMFBWAIGRX-UHFFFAOYSA-N
Synonym 4-tert-butoxycarbonyl amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2-carboxylic acid, boc-py-oh, 4-boc-amino-1-methylpyrrole-2-carboxylic acid, 4-tert-butoxycarbonyl amino-1-methylpyrrole-2-carboxylic acid, 4-boc-amino-1-methyl-1h-pyrrole-2-carboxylic acid, 4-tert-butoxycarbonylamino-1-methyl-1h-pyrrole-2 carboxylic acid, boc-4-amino-1-methylpyrrole-2-carboxylic acid
PubChem CID 2735636
IUPAC Name 1-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]pyrrole-2-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1=CN(C(=C1)C(=O)O)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 240.26
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butoxycarbonyl)amino]-1-methyl-1H-pyrrole-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.