missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Amylphenol, 99%

Product Code. 10193840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Unit Size:
100g
250g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193840 100 g 100g
10061910 5 g 5g
10438910 250 g 250g
10666562 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10193840 Supplier Thermo Scientific Acros Supplier No. 167331000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-46-6 | C11H16O | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-46-6
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00002369
InChI Key NRZWYNLTFLDQQX-UHFFFAOYSA-N
Synonym 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol
PubChem CID 6643
ChEBI CHEBI:35096
IUPAC Name 4-(2-methylbutan-2-yl)phenol
SMILES CCC(C)(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >91.0°C
Color White to Yellow
Boiling Point 255.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5C(CH3)2C6H4OH
Quantity 100 g
Beilstein 06, 548
Merck Index 15, 7255
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, benzene and chloroform
Viscosity 2.9 mPa.s (100°C)
Formula Weight 164.25
Percent Purity 99%
Physical Form Briquettes or Flakes to Coarse Powder
Chemical Name or Material 4-tert-Amylphenol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Amylphenol
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.