missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfonic calix[4]arene hydrate, 95%, Thermo Scientific Chemicals

Product Code. 10484735
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C28H24O16S4·xH2O
Molecular Weight (g/mol) 744.75
MDL Number MFCD16495912

Specifications

Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol
Formula Weight 744.75
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Sulfonic calix[4]arene hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'