missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfobenzoic acid monopotassium salt, 95%

Product Code. p-7024558
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11484017

Brand: Thermo Scientific Alfa Aesar A19432.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5399-63-3 | C7H5KO5S | 240.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5399-63-3
Molecular Formula C7H5KO5S
Molecular Weight (g/mol) 240.27
MDL Number MFCD00007509
InChI Key PXRJBUPXKDXDLG-UHFFFAOYSA-M
Synonym benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
PubChem CID 23670833
IUPAC Name potassium;4-sulfobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Formula Weight 240.28
Percent Purity 95%
Chemical Name or Material 4-Sulfobenzoic acid monopotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.