Learn More
Hanna Instruments™ 4-Ring Interchangeable Conductivity Probe
Product Code. 11537799
Change view
Click to view available options
Length (Metric) Cable:
1 m
Unit Size:
Each
Product Code. Length (Metric) Cable unitSize
11537799 1 m Each
1 options
Product Code. 11537799 Supplier Hanna Instruments™ Supplier No. HI76301D
Only null left
Add to basket
Add to basket
Specifications
Connector Types DIN
For Use With (Equipment) HI 8633 and HI 8734 Meter
Type 4-Ring Interchangeable Conductivity Probe
Length (Metric) Cable 1 m
Probe Conductivity