Learn More
Hanna Instruments™ 4-Ring Conductivity Probes
Product Code. 11507809
Change view
Click to view available options
For Use With (Equipment):
HI 8733
HI 8733, HI 9033, HI 9034, HI 87314
HI 9835, HI 98360
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11507809 HI 9835, HI 98360 Each
10090761 HI 8733, HI 9033, HI 9034, HI 87314 Each
10777151 HI 8733 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11507809 Supplier Hanna Instruments™ Supplier No. HI76309
  • ONLINE EXCLUSIVE PRICE
    £572.54
    £461.41 / Pack of 1
    Save £111.13
    19% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Connector Types DIN
For Use With (Application) Measures electrical conductivity in liquids
For Use With (Equipment) HI 9835, HI 98360
Length (Metric) Cable 1 m
Temperature Sensor Yes