missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™

Product Code. 10410584
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10410584 10 g 10g
10653793 250 mg 250mg
10733253 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10410584 Supplier Thermo Scientific Maybridge Supplier No. MO00517EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 165669-32-9
Molecular Formula C10H12ClNO4S2
Molecular Weight (g/mol) 309.779
InChI Key ZXYZKIFFWKXROK-UHFFFAOYSA-N
Synonym 4-pyrrolidine-1-sulfonyl-benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzene-1-sulfonyl chloride, 4-pyrrolidin-1-ylsulfonyl benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzenesulfonyl chloride, benzenesulfonylchloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulphonyl benzenesulphonyl chloride, 4-pyrrolidine-1-sulfonyl phenylsulfonyl chloride, benzenesulfonyl chloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulfonyl benzene-1-sulfonyl chloride
PubChem CID 2779793
IUPAC Name 4-pyrrolidin-1-ylsulfonylbenzenesulfonyl chloride
SMILES C1CCN(C1)S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 309.79
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.