missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10205944
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10205944 10 g 10g
10295934 250 mg 250mg
10441694 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205944 Supplier Thermo Scientific Maybridge Supplier No. CC55601EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159691-25-5
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
InChI Key DOEIPZUTRVMWAB-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-ylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoic acid, 4-pyrrolidin-1-yl methyl benzoic acid, 4-pyrrolidinylmethyl benzoic acid, 4-pyrrolidin-1-ylmethyl-benzoicacid, 4-1-pyrrolidinylmethyl-benzoic acid, benzoic acid, 4-1-pyrrolidinylmethyl-, hydrochloride
PubChem CID 1092976
IUPAC Name 4-(pyrrolidin-1-ylmethyl)benzoic acid
SMILES C1CCN(C1)CC2=CC=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.