missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10336384
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10336384 10 g 10g
10633245 250 mg 250mg
10183224 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10336384 Supplier Thermo Scientific Maybridge Supplier No. CC55614EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 142335-64-6
Molecular Formula C11H17N2
Molecular Weight (g/mol) 177.27
MDL Number MFCD03724739
InChI Key SFEAIUCOZWDYMJ-UHFFFAOYSA-O
Synonym 4-pyrrolidin-1-ylmethyl aniline, 4-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 4-1-pyrrolidinylmethyl, 4-1-pyrrolidinylmethyl aniline, 4-pyrrolidin-1-ylmethyl-aniline, 4-pyrrolidinylmethyl phenylamine, 1-4-aminobenzyl pyrrolidine, 4-pyrrolizinomethyl aniline, 4-pyrrolidinyl-methyl aniline, cbi-bb zero/006543
PubChem CID 776852
IUPAC Name 4-(pyrrolidin-1-ylmethyl)aniline
SMILES NC1=CC=C(C[NH+]2CCCC2)C=C1

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 176.26
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.