missing translation for 'onlineSavingsMsg'
Learn More

(4-Pyrimidin-5-ylphenyl)methanol, ≥97%, Thermo Scientific™

£19.60 - £49.55

Chemical Identifiers

CAS 198084-13-8
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD06802874
InChI Key QPOIDCHBWLGFGU-UHFFFAOYSA-N
Synonym 4-pyrimidin-5-ylphenyl methanol, 4-pyrimidin-5-yl phenyl methanol, 4-5-pyrimidinyl phenyl methanol, 4-pyrimidin-5-yl-phenyl-methanol, 4-5-pyrimidinyl-benzyl alcohol, benzenemethanol,4-5-pyrimidinyl, 4-pyrimidin-5-ylphenyl methan-1-ol, swx
PubChem CID 7162046
IUPAC Name (4-pyrimidin-5-ylphenyl)methanol
SMILES C1=CC(=CC=C1CO)C2=CN=CN=C2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10056142
View Documents Promotion Details
Thermo Scientific Maybridge
CC44309CB
250 mg Amber Glass Bottle
£19.60
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10413793
View Documents Promotion Details
Thermo Scientific Maybridge
CC44309DA
1 g Amber Glass Bottle
£49.55
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

198084-13-8
186.214
QPOIDCHBWLGFGU-UHFFFAOYSA-N
7162046
C1=CC(=CC=C1CO)C2=CN=CN=C2
C11H10N2O
MFCD06802874
4-pyrimidin-5-ylphenyl methanol, 4-pyrimidin-5-yl phenyl methanol, 4-5-pyrimidinyl phenyl methanol, 4-pyrimidin-5-yl-phenyl-methanol, 4-5-pyrimidinyl-benzyl alcohol, benzenemethanol,4-5-pyrimidinyl, 4-pyrimidin-5-ylphenyl methan-1-ol, swx
(4-pyrimidin-5-ylphenyl)methanol

Specifications

198084-13-8
100.0
C11H10N2O
4-pyrimidin-5-ylphenyl methanol, 4-pyrimidin-5-yl phenyl methanol, 4-5-pyrimidinyl phenyl methanol, 4-pyrimidin-5-yl-phenyl-methanol, 4-5-pyrimidinyl-benzyl alcohol, benzenemethanol,4-5-pyrimidinyl, 4-pyrimidin-5-ylphenyl methan-1-ol, swx
C1=CC(=CC=C1CO)C2=CN=CN=C2
186.214
186.21
(4-Pyrimidin-5-ylphenyl)methanol
96.5
97%
MFCD06802874
QPOIDCHBWLGFGU-UHFFFAOYSA-N
(4-pyrimidin-5-ylphenyl)methanol
7162046
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications