Learn More
4-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™
£33.40 - £37.00
Chemical Identifiers
CAS | 216959-91-0 |
---|---|
Molecular Formula | C11H8N2O2 |
Molecular Weight (g/mol) | 200.197 |
MDL Number | MFCD05864811 |
InChI Key | CCBKADQVSXLSDN-UHFFFAOYSA-N |
Synonym | 4-pyrimidin-5-yl benzoic acid, 4-pyrimidin-5-yl-benzoic acid, 4-5-pyrimidinyl benzoic acid, benzoic acid, 4-5-pyrimidinyl |
PubChem CID | 4187247 |
IUPAC Name | 4-pyrimidin-5-ylbenzoic acid |
SMILES | C1=CC(=CC=C1C2=CN=CN=C2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10670695
|
Thermo Scientific Maybridge
CC44301CB |
250 mg | Amber Glass Bottle |
£33.40
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219662
|
Thermo Scientific Maybridge
CC44301DA |
1 g | Amber Glass Bottle | ||||||
Description
Chemical Identifiers
216959-91-0 | |
200.197 | |
CCBKADQVSXLSDN-UHFFFAOYSA-N | |
4187247 | |
C1=CC(=CC=C1C2=CN=CN=C2)C(=O)O |
C11H8N2O2 | |
MFCD05864811 | |
4-pyrimidin-5-yl benzoic acid, 4-pyrimidin-5-yl-benzoic acid, 4-5-pyrimidinyl benzoic acid, benzoic acid, 4-5-pyrimidinyl | |
4-pyrimidin-5-ylbenzoic acid |
Specifications
216959-91-0 | |
100.0 | |
C11H8N2O2 | |
4-pyrimidin-5-yl benzoic acid, 4-pyrimidin-5-yl-benzoic acid, 4-5-pyrimidinyl benzoic acid, benzoic acid, 4-5-pyrimidinyl | |
C1=CC(=CC=C1C2=CN=CN=C2)C(=O)O | |
200.197 | |
200.2 | |
4-Pyrimidin-5-ylbenzoic acid |
96.5 | |
97% | |
MFCD05864811 | |
CCBKADQVSXLSDN-UHFFFAOYSA-N | |
4-pyrimidin-5-ylbenzoic acid | |
4187247 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.