missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzonitrile, 95%, Thermo Scientific™

Product Code. 10217153
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217153 1 g Amber Glass Bottle 1g
10692855 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217153 Supplier Thermo Scientific Maybridge Supplier No. CC56216DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 78322-96-0
Molecular Formula C11H7N3
Molecular Weight (g/mol) 181.198
MDL Number MFCD09817491
InChI Key WGCKVXHOTDDNOA-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzonitrile, 4-cyanophenyl pyrimidine, 2-4-cyanophenyl pyrimidine, 4-pyrimidin-2-yl-benzonitrile, 4-2-pyrimidinyl benzonitrile, benzonitrile, 4-2-pyrimidinyl, 4-pyrimidin-2-ylbenzenecarbonitrile
PubChem CID 15486310
IUPAC Name 4-pyrimidin-2-ylbenzonitrile
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.2
Percent Purity 95%
Chemical Name or Material 4-Pyrimidin-2-ylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.