Learn More
4-Pyrimidin-2-ylbenzaldehyde, 95%, Maybridge™
Brand: Maybridge CC56204DE
Code : 31
Additional Details : CAS Number : 77232-38-3 Weight : 0.05500kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
77232-38-3 | |
184.198 | |
JUYLMPWKVWDXBE-UHFFFAOYSA-N | |
12703722 | |
C1=CN=C(N=C1)C2=CC=C(C=C2)C=O |
C11H8N2O | |
MFCD08056282 | |
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl | |
4-pyrimidin-2-ylbenzaldehyde |
Specifications
4-Pyrimidin-2-ylbenzaldehyde | |
5g | |
94.5 | |
95% | |
MFCD08056282 | |
JUYLMPWKVWDXBE-UHFFFAOYSA-N | |
4-pyrimidin-2-ylbenzaldehyde | |
12703722 | |
95% |
Amber Glass Bottle | |
77232-38-3 | |
100.0 | |
C11H8N2O | |
4-pyrimidin-2-yl benzaldehyde, 4-2-pyrimidinyl benzaldehyde, 2-4-formylphenyl pyrimidine, 4-pyrimidin-2-yl-benzaldehyde, 4-2-pyrimidinyl-benzaldehyde, benzaldehyde,4-2-pyrimidinyl, benzaldehyde, 4-2-pyrimidinyl | |
C1=CN=C(N=C1)C2=CC=C(C=C2)C=O | |
184.198 | |
184.2 |