missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinemethanol, 99%

Product Code. 11477706
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477706 25 g 25g
11487706 100 g 100g
11497706 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477706 Supplier Thermo Scientific Alfa Aesar Supplier No. A14698.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-95-8 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-95-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006442
InChI Key PTMBWNZJOQBTBK-UHFFFAOYSA-N
Synonym 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol
PubChem CID 11472
SMILES OCC1=CC=NC=C1

Specifications

Melting Point 53°C to 55°C
Boiling Point 141°C (12 mmHg)
Flash Point 148°C (298°F)
Quantity 25 g
Beilstein 107850
Sensitivity Hygroscopic
IUPAC Name pyridin-4-ylmethanol
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 4-Pyridinemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.