missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetic acid hydrochloride, 98+%

Product Code. 11486907
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486907 25 g 25g
11493990 1 g 1g
11476907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486907 Supplier Thermo Scientific Alfa Aesar Supplier No. A14348.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6622-91-9 | C7H8ClNO2 | 173.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-91-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012827
InChI Key WKJRYVOTVRPAFN-UHFFFAOYSA-N
Synonym 4-pyridineacetic acid hydrochloride, 4-pyridylacetic acid hydrochloride, 2-pyridin-4-yl acetic acid hydrochloride, 4-carboxymethylpyridinium chloride, 4-acetic acid pyridine hydrochloride, 4-pyridineacetic acid hcl, 4-pyridine acetic acid hydrochloride, 2-4-pyridyl acetic acid hydrochloride, 4-acetylpyridine hydrochloride, 4-pyridineacetic acid, hydrochloride
PubChem CID 81097
IUPAC Name 2-pyridin-4-ylacetic acid;hydrochloride
SMILES C1=CN=CC=C1CC(=O)O.Cl

Specifications

Melting Point ∼136°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 3696648
Formula Weight 173.6
Percent Purity ≥98%
Chemical Name or Material 4-Pyridineacetic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.