missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridin-4-ylbenzaldehyde, 97%, Thermo Scientific™

Product Code. 10046212
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10046212 Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046212 Supplier Thermo Scientific Maybridge Supplier No. CC35404CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 99163-12-9
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H9NO
Quantity 250 mg
Synonym 4-pyridin-4-yl-benzaldehyde, 4-pyridin-4-yl benzaldehyde, 4-4-pyridyl benzaldehyde, 4-4-pyridinyl benzaldehyde, 4-4-formylphenyl pyridine, akos bar-0607, benzaldehyde, 4-4-pyridinyl, acmc-209sbt
InChI Key MBJXDIYHLGBQOT-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C=O)C2=CC=NC=C2
IUPAC Name 4-pyridin-4-ylbenzaldehyde
Molecular Weight (g/mol) 183.21
PubChem CID 639673
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-Pyridin-4-ylbenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.