missing translation for 'onlineSavingsMsg'
Learn More

(4-Pyrid-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10178002
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178002 1 g 1g
10527683 250 mg 250mg
10377404 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10178002 Supplier Thermo Scientific Maybridge Supplier No. CC40009DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 217189-04-3
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD04116293
InChI Key ZHIJCVCCKVZBHE-UHFFFAOYSA-N
Synonym 4-pyrid-3-ylphenyl methanol, 4-pyridin-3-yl phenyl methanol, 4-pyridin-3-ylphenyl methanol, benzenemethanol,4-3-pyridinyl, 4-pyridin-3-yl-phenyl-methanol, 4-3-pyridyl-phenyl-methanol, 4-3-pyridyl phenyl methan-1-ol, 3-4-hydroxymethyl phenyl pyridine, 3-4-hydroxymethylphenyl pyridine
PubChem CID 1515243
IUPAC Name (4-pyridin-3-ylphenyl)methanol
SMILES C1=CC(=CN=C1)C2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (4-Pyrid-3-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.