missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 199.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Pyrid-2-yloxy)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'