missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrazolecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10376684
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
37718-11-9
C4H4N2O2
112.09
MFCD00011558
IMBBXSASDSZJSX-UHFFFAOYSA-N
4-pyrazolecarboxylic acid, 4-carboxypyrazole, pyrazole-4-carboxylic acid, unii-4xjz84gv8e, 1h-pyrazole-4-carboxylicacid, 4-carboxy-1h-pyrazole, 4xjz84gv8e, pyrazole-4-carboxylic, pubchem13293, pubchem22749
3015937
1H-pyrazole-4-carboxylic acid
C1=C(C=NN1)C(=O)O
This item is not returnable. View return policy
37718-11-9
C4H4N2O2
112.09
MFCD00011558
IMBBXSASDSZJSX-UHFFFAOYSA-N
4-pyrazolecarboxylic acid, 4-carboxypyrazole, pyrazole-4-carboxylic acid, unii-4xjz84gv8e, 1h-pyrazole-4-carboxylicacid, 4-carboxy-1h-pyrazole, 4xjz84gv8e, pyrazole-4-carboxylic, pubchem13293, pubchem22749
3015937
1H-pyrazole-4-carboxylic acid
C1=C(C=NN1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 282°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 112.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Pyrazolecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Pyrazolecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'