missing translation for 'onlineSavingsMsg'
Learn More

4-Propoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 10752463
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752463 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10752463 Supplier Thermo Scientific Maybridge Supplier No. BTB09860DA
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 60758-84-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H11NO
Linear Formula CH3(CH2)2OC6H4CN
MDL Number MFCD00173874
Quantity 1 g
Synonym 4-propoxy-benzonitrile, 4-propoxybenzenecarbonitrile, 4-n-propoxybenzonitrile, benzonitrile, 4-propoxy
InChI Key VYOPNUZYNSNYRA-UHFFFAOYSA-N
SMILES CCCOC1=CC=C(C=C1)C#N
IUPAC Name 4-propoxybenzonitrile
Molecular Weight (g/mol) 161.204
PubChem CID 322939
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 4-Propoxybenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.