Learn More
Labconco 4 place Microplate Rotor - CentriVap Pro
GREENER_CHOICE
Product Code. p-200132245 Shop All Labconco Corporation Products
Click to view available options
Unit Size:
Each
Material (Coating):
PTFE
Product Code. 30533406
Brand: Labconco 7555800
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stainless steel material
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Rotor System CentriVap Pro
Dimensions (L x W x H) 25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.)