Learn More
Labconco 4 place Microplate Rotor - CentriVap Pro
Product Code. 30533405 Shop All Labconco Corporation Products
Change view
Click to view available options
Material (Coating):
PTFE
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Coating) Unit Size
30533405 PTFE Each
30533406 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30533405 Supplier Labconco Supplier No. 7555801
  • ONLINE EXCLUSIVE PRICE
    £1940.00
    £1773.00 / Pack of 1
    Save £167.00
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Rotors for use with Labconco™ CentriVap™ Pro Vacuum Concentrators. Stainless steel and PTFE coated options available.
  • Compatible with CentriVap Pro Vacuum Concentrators
  • Stainless steel material
  • Optional PTFE coated
Specifications
Material Stainless Steel
Type Rotor
Material (Coating) PTFE
Rotor System CentriVap Pro
Dimensions (L x W x H) 25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.)